Actinomycin D 1 mg/ml in DMSO, sterile filtered

Supplier: Thermo Scientific Chemicals
Warning

J67160-8EQ J67160-XF
AAJ67160-8EQEA 619 USD
AAJ67160-8EQ AAJ67160-XF
Actinomycin D 1 mg/ml in DMSO, sterile filtered
Actinomycin D

Actinomycin D, 1 mg/ml in DMSO, sterile-filtered


  • Liquid


Actinomycin D has a wide-ranging applications ranging from a selection agent in cell culture, use in studies of suppressing HIV-replication and programmed cell death of PC12 cells. It acts as an antibiotic in treatment of various malignant neoplasms, including Wilms tumor and the sarcomas.


Caution: Research use only: Not intended for animal or human diagnostic or therapeutic use.

Formula: C₆₂H₈₆N₁₂O₁₆
Storage Temperature: Freezer
MDL Number: MFCD00005033
CAS Number: 50-76-0
EINECS: 200-063-6
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR