Carbenicillin disodium salt 50 mg/mL in distilled water, sterile filtered

Supplier: Thermo Scientific Chemicals
Danger

J67159-AD J67159-AE
AAJ67159-ADEA 333 USD
AAJ67159-AD AAJ67159-AE
Carbenicillin disodium salt 50 mg/mL in distilled water, sterile filtered
Carbenicillin disodium salt

Carbenicillin disodium salt, 50 mg/ml in distilled water, sterile-filtered

Formula: C₁₇H₁₆N₂O₆S·2Na
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS Number: 4800-94-6
EINECS: 225-360-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR