Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 80 nm OD1 (Abs. 553 nm)

Supplier: Thermo Scientific Chemicals

J67109-AC J67109-AE
AAJ67109-ACEA 94.7 USD
AAJ67109-AC AAJ67109-AE
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 80 nm OD1 (Abs. 553 nm)
Gold

Suitable for adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.


Liquid


Applications: Suitable for adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.

Formula: Au
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR