Silver 0.02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 100 nm (Abs. 490 nm)

Supplier: Thermo Scientific Chemicals

J67099-AC J67099-AE
AAJ67099-ACEA 90.5 USD
AAJ67099-AC AAJ67099-AE
Silver 0.02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 100 nm (Abs. 490 nm)
Silver

An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range. Readily binds proteins and other ligands for conjugate and sensor development.


Liquid


Applications: An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range. Readily binds proteins and other ligands for conjugate and sensor development.

Formula: Ag
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR