Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 100 nm OD1 (Abs. 572 nm)

Supplier: Thermo Scientific Chemicals

J67095-AC J67095-AE
AAJ67095-ACEA 96.1 USD
AAJ67095-AC AAJ67095-AE
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 100 nm OD1 (Abs. 572 nm)
Gold

Liquid


CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Red to pink MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR