Glycine, 0.2 M Buffer Solution, pH 2.5, Low Endotoxin, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J67094-AE
J67094-AK
AAJ67094-AEEA
86.08
USD
AAJ67094-AE
AAJ67094-AK
Glycine, 0.2 M Buffer Solution, pH 2.5, Low Endotoxin, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 56-40-6
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00008131
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...