Moxalactam sodium salt 50 mg/ml in distilled water, sterile filtered
Supplier: Thermo Scientific Chemicals
|
Danger
|
J67056-XF
AAJ67056-XFEA
70.7
USD
AAJ67056-XF
Moxalactam sodium salt 50 mg/ml in distilled water, sterile filtered
Moxalactam sodium salt
Concentration: 50 mg/ml
CAS No.: 64953-12-4
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD03427565
Formula:
C₂₀H₁₈N₆Na₂O₉S Storage Temperature: Refrigerator |
MDL Number:
MFCD03427565 CAS Number: 64953-12-4 EINECS: 265-288-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...