Gold amine functionalized, 3kDa PEGylated, nanoparticles 5 nm OD50 (Abs. 515-520 nm)
Supplier: Thermo Scientific Chemicals
J67053-AMI
J67053-XF
AAJ67053-AMIEA
298
USD
AAJ67053-AMI
AAJ67053-XF
Gold amine functionalized, 3kDa PEGylated, nanoparticles 5 nm OD50 (Abs. 515-520 nm)
Gold
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Red
MDL No.: MFCD00003436
Formula:
Au Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...