D-Cycloserine 20 mg/mL in distilled water, sterile filtered

Supplier: Thermo Scientific Chemicals

J67020-AC J67020-AD
AAJ67020-ACEA 166 USD
AAJ67020-AC AAJ67020-AD
D-Cycloserine 20 mg/mL in distilled water, sterile filtered
D-Cycloserine

D-Cycloserine, 20 mg/ml in distilled water, sterile-filtered

Formula: C₃H₆N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00005353
CAS Number: 68-41-7
EINECS: 200-688-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR