Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 40 nm OD1 (Abs. 530 nm)
Supplier: Thermo Scientific Chemicals
J67001-AC
J67001-AE
AAJ67001-ACEA
95.3
USD
AAJ67001-AC
AAJ67001-AE
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 40 nm OD1 (Abs. 530 nm)
Gold
Liquid
CAS No.: 7440-57-5
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Red to pink
Odor: Odorless
MDL No.: MFCD00003436
Formula:
Au Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...