Ampicillin 100 mg/ml in aqueous solution ready-to-use
Supplier: Thermo Scientific Chemicals
|
Danger
|
Filtered through a 0.2 µm filter.
J66972-AB
J66972-AC
AAJ66972-ABEA
182.36
USD
AAJ66972-AB
AAJ66972-AC
Ampicillin 100 mg/ml in aqueous solution ready-to-use
Ampicillin
Pale yellow liquid
- Light sensitive
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Sensitivity: Light Sensitive
Appearance: Pale yellow
Odor: Odorless
Formula:
C₁₆H₁₉N₃O₄S Storage Temperature: Refrigerator |
MDL Number:
MFCD00005175 CAS Number: 69-53-4 EINECS: 200-708-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...