Substance P,Biotin
Supplier: Thermo Scientific Chemicals
J66653-LB0
J66653-MA
AAJ66653-LB0EA
337
USD
AAJ66653-LB0
AAJ66653-MA
Substance P,Biotin
Proteins and Peptides
Biotin-Substance P
CAS No.: 87468-58-4
Molecular Formula: C73H112N20O15S2
Formula Weight: 1573.94
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00143749
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...