Riboflavine (vitamin B2) 97% for electrophoresis
Supplier: Thermo Scientific Chemicals
Synonyms:
Vitamin B2, Lactoflavin, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione, (-)-Riboflavin, Vitamine G
J66632-14
AAJ66632-14EA
62.8
USD
AAJ66632-14
Riboflavine (vitamin B2) 97% for electrophoresis
Riboflavine (Vitamin B2)
Powder
Purity: 97%
CAS No.: 83-88-5
Molecular Formula: C17H20N4O6
Formula Weight: 376.37
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Orange-yellow
Odor: Odorless
Melting Point: 290°
MDL No.: MFCD00005022
Formula:
C₁₇H₂₀N₄O₆ MW: 376.37 g/mol Melting Pt: 290 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005022 CAS Number: 83-88-5 EINECS: 201-507-1 Merck Index: 14,8200 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...