DAPT

Supplier: Thermo Scientific Chemicals

Synonyms: N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester, tert-Butyl (S)-2-((S)-2-(2-(3,5-difluorophenyl)acetamido)propanamido)-2-phenylacetate, (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu, (3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester

J65864-MA
AAJ65864-MAEA 232 USD
AAJ65864-MA
DAPT
DAPT

A cell-permeable dipeptide that suppresses A_x001A_ production by blocking _x001A_-secretase


Lyophilized


Applications: A cell-permeable dipeptide that suppresses A production by blocking -secretase

Formula: C₂₃H₂₆F₂N₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD04974585
CAS Number: 208255-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR