Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.9%, Ultrapure

Supplier: Thermo Scientific Chemicals
Warning

J65594-A1 J65594-A7
AAJ65594-A1EA 166.96 USD
AAJ65594-A1 AAJ65594-A7
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.9%, Ultrapure
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

Crystalline powder


CAS: 77-86-1 MDL: MFCD00004679

Formula: NH₂C(CH₂OH)₃
Boiling Pt: 219…220 °C
Melting Pt: 167…172 °C
Density: 1.35
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 14,9772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Minimum 99.9


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR