EDTA (ethylenediamine tetraacetic acid) ≥99.4% for electrophoresis

Supplier: Thermo Scientific Chemicals
Warning

J65585-22 J65585-36 J65585-A4
AAJ65585-22EA 33.28 USD
AAJ65585-22 AAJ65585-36 AAJ65585-A4
EDTA (ethylenediamine tetraacetic acid) ≥99.4% for electrophoresis
EDTA (Ethylenediamine tetraacetic acid)

Powder


Purity: 99.4+% CAS No.: 60-00-4 Molecular Formula: C10H16N2O8 Formula Weight: 292.24 Physical Form: Powder Density: 0.68 Appearance: White Melting Point: ca 245° dec. Beilstein Reference: 1716295 Merck Reference: 14,3517 MDL No.: MFCD00003541

Formula: C₁₀H₁₆N₂O₈
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: ∼ 245 °C (dec.)
Density: 0.68
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 14,3517
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR