Arg-Gly-Asp
Supplier: Thermo Scientific Chemicals
J65468-LB0
AAJ65468-LB0EA 160 USD
AAJ65468-LB0
Arg-Gly-Asp
Proteins and Peptides
Fragment from cell-attachment domain of fibronectin; a cell-attachment peptide.
  • Soluble in 0.1 N acetic acid at 20 mg/mL
  • Synonyms: Arginyl-glycyl-aspartic acid; RGD