2'-Deoxy-5-methylcytidine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one, 5-mdC, 5-Methyl-2'-deoxycytidine, 1-(2-Deoxy-beta-D-ribofuranosyl)-5-methylcytosine

J65209-03
AAJ65209-03EA 135.02 USD
AAJ65209-03
2'-Deoxy-5-methylcytidine 99%
2'-Deoxy-5-methylcytidine

Powder


Purity: 99% CAS No.: 838-07-3 Molecular Formula: C10H15N3O4 Formula Weight: 241.24 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00006549

Formula: C₁₀H₁₅N₃O₄
MW: 241.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006549
CAS Number: 838-07-3
EINECS: 212-655-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR