1-Aminocyclopropanecarboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Amino-1-cyclopropanecarboxylic acid
J65118-03
J65118-MD
AAJ65118-03EA
173
USD
AAJ65118-03
AAJ65118-MD
1-Aminocyclopropanecarboxylic acid 98%
1-Aminocyclopropanecarboxylic acid
A NMDA glutamate receptor agonist at glycine site
Crystalline powder
Formula:
C₄H₇NO₂ MW: 101.11 g/mol Melting Pt: 229…231 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009944 CAS Number: 22059-21-8 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...