3-Isobutyl-1-methylxanthine
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,6-Dihydroxy-3-isobutyl-1-methylpurine, IBMX, 3,7-Dihydro-3-isobutyl-1-methyl-1H-purine-2,6-dione, 3-Isobutyl-1-methyl-3,7-dihydro-1H-purine-2,6-dione
J64598-MC
AAJ64598-MCEA
57.55
USD
AAJ64598-MC
3-Isobutyl-1-methylxanthine
3-Isobutyl-1-methylxanthine
Solid
CAS No.: 28822-58-4
Molecular Formula: C10H14N4O2
Formula Weight: 222.24
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 200-201°
MDL No.: MFCD00005584
Formula:
C₁₀H₁₄N₄O₂ Melting Pt: 200…201 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005584 CAS Number: 28822-58-4 EINECS: 249-259-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...