2',3'-Dideoxyadenosine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
((2S,5R)-5-(6-Amino-9H-purin-9-yl)tetrahydrofuran-2-yl)methanol
J64573-MB
AAJ64573-MBEA
110
USD
AAJ64573-MB
AAJ64573-MC
AAJ64573-MF
2',3'-Dideoxyadenosine 98%
2',3'-Dideoxyadenosine
Powder
Purity: 98%
CAS No.: 4097-22-7
Molecular Formula: C10H13N5O2
Formula Weight: 235.24
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Melting Point: 181-184°
Beilstein Reference: 619924
Merck Reference: 14,3101
MDL No.: MFCD00010534
Formula:
C₁₀H₁₃N₅O₂ MW: 235.24 g/mol Melting Pt: 181…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010534 CAS Number: 4097-22-7 EINECS: 223-853-2 Merck Index: 14,3101 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...