Flt-3 inhibitor

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(3,4-dimethoxybenzamido)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide, TCS 359, TCS-359

J64484-MA
AAJ64484-MAEA 272 USD
AAJ64484-MA AAJ64484-MF
Flt-3 inhibitor
Flt-3 inhibitor

A cell-permeable thienylcarboxamide compound that acts as a potent, ATP-competitive, and highly selective Flt-3 inhibitor


  • Light Sensitive


Powder

Formula: C₁₈H₂₀N₂O₄S
MW: 360.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00617269
CAS Number: 301305-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR