Chloroquine diphosphate salt 98%

Supplier: Thermo Scientific Chemicals
Warning

J64459-22 J64459-14 J64459-18 J64459-30
AAJ64459-22EA 277 USD
AAJ64459-22 AAJ64459-14 AAJ64459-18 AAJ64459-30
Chloroquine diphosphate salt 98%
Chloroquine diphosphate salt

A mild immunosupressant


Purity: 98% CAS No.: 50-63-5 Molecular Formula: C18H26ClN3.2H3PO4 Formula Weight: 515.87 Physical Form: Powder Appearance: White to off-white Melting Point: 213-216° Solubility: Soluble in water Beilstein Reference: 4223142 MDL No.: MFCD00069852


Applications: A mild immunosupressant

Formula: C₁₈H₂₆ClN₃
Melting Pt: 213…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00069852
CAS Number: 50-63-5
EINECS: 200-055-2
Merck Index: 14,2163

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR