5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) 98%

Supplier: Thermo Scientific Chemicals

J64451-03 J64451-MD
AAJ64451-03EA 284 USD
AAJ64451-03 AAJ64451-MD
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) 98%
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)

Substrate for alkaline phosphatase


Powder

Formula: C₈H₄BrClNNa₂O₄P
Storage Temperature: Refrigerator
MDL Number: MFCD00036757
CAS Number: 102185-33-1

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR