Sodium dodecyl sulphate (SDS) 97% for electrophoresis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
Powder
Purity: 98%
CAS No.: 151-21-3
Molecular Formula: C12H25NaO4S
Formula Weight: 288.38
Physical Form: Powder
Appearance: White
Melting Point: ca 206°
Solubility: Soluble in water
Beilstein Reference: 3599286
Merck Reference: 14,8636
UN No.: 2926
MDL No.: MFCD00036175
Formula:
CH₃(CH₂)₁₁OSO₃Na Melting Pt: ∼ 206 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: ≥ 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1,III Merck Index: 14,8636 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...