2-Methyl-5-hydroxytryptamine hydrochloride 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Me-5HT, 3-(2-aminoethyl)-2-methyl-1H-indol-5-ol hydrochloride

J64203-MB
AAJ64203-MBEA 276 USD
AAJ64203-MB
2-Methyl-5-hydroxytryptamine hydrochloride 97%
2-Methyl-5-hydroxytryptamine hydrochloride
Formula: C₁₁H₁₄N₂O·HCl
MW: 244.73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02677991
CAS Number: 845861-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR