N-Hydroxy-N'-phenyloctanediamide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N1-Hydroxy-N8-phenyloctanediamide, Suberoylanilide Hydroxamic Acid, Vorinostat

J64198-MC
AAJ64198-MCEA 430.47 USD
AAJ64198-MC
N-Hydroxy-N'-phenyloctanediamide 98%
N-Hydroxy-N'-phenyloctanediamide
Formula: C₁₄H₂₀N₂O₃
MW: 264.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00945317
CAS Number: 149647-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR