Factor XA protease digestion buffer, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63878-AK
AAJ63878-AKEA
66.1
USD
AAJ63878-AK
Factor XA protease digestion buffer, Thermo Scientific
Buffers
Enzyme Buffers
Liquid
Physical Form: Liquid
Applications: For His-tag protein purification from mammalian cells
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...