N,N,N',N'-Tetramethylethylenediamine (TEMED) for electrophoresis

Supplier: Thermo Scientific Chemicals
Danger

J63734-AC J63734-AE
AAJ63734-ACEA 71.34 USD
AAJ63734-AC AAJ63734-AE
N,N,N',N'-Tetramethylethylenediamine (TEMED) for electrophoresis
N,N,N',N'-Tetramethylethylenediamine (TEMED)

Liquid


CAS: 110-18-9 MDL: MFCD00008335

Formula: C₆H₁₆N₂
Boiling Pt: 120…122 °C
Melting Pt: –55 °C
Density: 0.775
Flash Pt: 18 °C (154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008335
CAS Number: 110-18-9
EINECS: 203-744-6
UN: 2372
ADR: 3,II
Merck Index: 14,9134
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6838f1f5b7d0f69dcc81a8ed247f6c178dad231c&vid=ea361367e9944e4101e38622fdd18b07756eb676&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR