CHES, 0.5 M Buffer Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63670-AE
J63670-AK
AAJ61492-AEEA
68.8
USD
AAJ61492-AE
AAJ61492-AK
AAJ62333-AE
AAJ62333-AK
AAJ63670-AE
AAJ63670-AK
CHES, 0.5 M Buffer Solutions, Thermo Scientific
Buffers
Biological Buffers
2-(Cyclohexylamino)ethanesulfonic acid, CHES.
CAS No.: 103-47-9
MDL No.: MFCD00003835
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...