D-Sphingosine ≥99%
Supplier: Thermo Scientific Chemicals
Synonyms:
D-Sphingosine, trans-D-erythro-2-Amino-4-octadecene-1,3-diol, (2S,3R,E)-2-Amino-4-octadecene-1,3-diol, 4-Sphingenine, (2S,3R,E)-2-Aminooctadec-4-ene-1,3-diol
D-erythro-Sphingosine is used to inhibit protein kinase C and calmodulin-dependent enzymes, induces apoptosis. And also used to constituent of cell membranes.
Purity: 99+%
CAS No.: 123-78-4
Molecular Formula: C18H37NO2
Formula Weight: 299.50
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 1727294
MDL No.: MFCD00036751
Applications: Inhibits protein kinase C and calmodulin-dependent enzymes. Induces apoptosis. A constituent of cell membranes
Caution: Store at –20 °C.
Formula:
C₁₈H₃₇NO₂ MW: 299.50 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00036751 CAS Number: 123-78-4 EINECS: 204-651-3 Merck Index: 14,8747 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...