Benzyldimethyltetradecylammonium chloride hydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

J63465-14
AAJ63465-14EA 47.5 USD
AAJ63465-14
Benzyldimethyltetradecylammonium chloride hydrate 98%
Benzyldimethyltetradecylammonium chloride hydrate

Phase transfer catalyst


Purity: 98% CAS No.: 139-08-2 Molecular Formula: C23H42ClN.xH2O Formula Weight: 368.05(anhy) Physical Form: Powder Appearance: White Beilstein Reference: 4062599 Merck Reference: 14,1059 UN No.: 3261 MDL No.: MFCD00011771


Phase transfer catalyst

Formula: C₂₃H₄₂ClN·H₂O
MW: 368.05 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00011771
CAS Number: 139-08-2
EINECS: 205-352-0
UN: 2923
ADR: 8,III
Merck Index: 14,1059

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR