BCA Protein Assay Kit, Thermo Scientific
Supplier: Thermo Scientific Chemicals
BCA Protein Assay Kit can be used to measure very dilute protein concentrations in very small sample volumes.
- Contains two separate solutions for estimation of protein concentration
- Form: Liquid
- Storage: Room temperature
Accurately measures protein concentrations from 0.5 to 30 µg/ml in tube assays and 1 to 20 µg/ml in 96- or 384-well plate assays. It is used for studying protein, that is protein interactions, measuring column fractions after affinity chromatography, estimating percent recovery of membrane proteins from cell extracts and high-throughput screening of fusion proteins.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...