Genistein ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone
4',5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.
Purity: 99+%
CAS No.: 446-72-0
Molecular Formula: C15H10O5
Formula Weight: 270.24
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Melting Point: ca 300°
Beilstein Reference: 263823
Merck Reference: 14,4391
MDL No.: MFCD00016952
Inhibitor of tyrosine protein kinase
Formula:
C₁₅H₁₀O₅ Melting Pt: ca 300 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00016952 CAS Number: 446-72-0 EINECS: 207-174-9 Merck Index: 14,4391 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...