Thrombin Buffer, pH 8.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63240-AP
AAJ63240-AKEA
35.58
USD
AAJ63240-AK
AAJ63240-AP
Thrombin Buffer, pH 8.0, Thermo Scientific
Buffers
Cloning Buffers
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...