Saponin Permeating Solution, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63209-AK
AAJ63209-AKEA 228.55 USD
AAJ63209-AK
Saponin Permeating Solution, Thermo Scientific
Electrophoresis Reagents Western Blotting Reagents
Saponin permeating solution, is used as a cell permeabilizing agent.
Saponin permeating solution is made by mixing and dissolving 5 g of saponin with 100 ml of 5X PBS (pH 7.4) followed by submicron filtration and/or autoclaving at 15 PSI for 60 minutes.