Calcium chloride 1 M in aqueous solution

Supplier: Thermo Scientific Chemicals
Warning

J63122-AD J63122-AE
AAJ63122-ADEA 93.63 USD
AAJ63122-AD AAJ63122-AE
Calcium chloride 1 M in aqueous solution
Calcium chloride

Liquid


CAS No.: 10043-52-4 Molecular Formula: CaCl2 Formula Weight: 110.99 Physical Form: Liquid Appearance: Colorless Odor: Odorless MDL No.: MFCD00010903

Formula: CaCl₂
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR