Buffer Kit for Antibody Purification on Protein A Column, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Liquid
Buffers to be used when performing antibody
Physical Form: Liquid
Applications: Buffers to be used when performing antibody purification using immobilized Protein A.
Delivery information: This kit contains: 1 L of 1 M Tris-HCl (pH 8.0) and 500 mL of 100 mM Glycine (pH 3.0).
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...