Anisomycin 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)pyrrolidin-3-yl acetate

J62964-MF
AAJ62964-MFEA 263 USD
AAJ62964-MF
Anisomycin 97%
Anisomycin

A protein synthesis inhibitor. Causes apoptosis of PC12 Cells


Powder

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
Melting Pt: 140…141 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077650
CAS Number: 22862-76-6
EINECS: 245-269-7
UN: 3462
ADR: "6.1",III
Merck Index: 14,670

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR