Destaining Bags, Ethidium Bromide, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62931-8U
AAJ62931-8UEA 219.86 USD
AAJ62931-8U
Destaining Bags, Ethidium Bromide, Thermo Scientific
Electrophoresis Reagents Stain Removal Reagents
Ethidium bromide de-staining bags with activation solution can be used to safely remove ethidium bromide from solution during overnight treatment.
  • Destain overnight with gentle rocking or mixing
  • Soluble in water
  • Stored at ambient temperature
Each bag extracts up to 5 mg of ethidium bromide from solution thus minimizing the exposure of research personnel to this material. The special absorbent mixture retains dye molecules in a convenient bag for removal and incineration. It provides a safer, more economical means for disposal of hazardous waste.