Chelerythrine chloride ≥99%

Supplier: Thermo Scientific Chemicals
Warning

J62906-LB0 J62906-MA J62906-MB
AAJ62906-LB0EA 143 USD
AAJ62906-LB0 AAJ62906-MA AAJ62906-MB
Chelerythrine chloride ≥99%
Chelerythrine chloride

A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells


Purity: 99+% CAS No.: 3895-92-9 Molecular Formula: C21H18ClNO4 Formula Weight: 383.82 Storage Temperature: -30°C to -10°C Physical Form: Solid Solubility: Soluble in DMSO at 2mg/ml Merck Reference: 14,2051 MDL No.: MFCD00060717


A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells

Formula: C₂₁H₁₈ClNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00060717
CAS Number: 3895-92-9
EINECS: 223-444-9
Merck Index: 14,2051

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR