Calcium chloride 100 mM in aqueous solution, sterile-filtered
Supplier: Thermo Scientific Chemicals
Synonyms:
Chloro calcium, Calcium dichloride, CaCl2
J62905-AP
J62905-K2
AAJ62905-APEA
158.32
USD
AAJ62905-AP
AAJ62905-K2
Calcium chloride 100 mM in aqueous solution, sterile-filtered
Calcium chloride
Liquid
CAS No.: 10043-52-4
Molecular Formula: CaCl2
Formula Weight: 110.99
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Formula:
CaCl₂ Storage Temperature: Ambient |
MDL Number:
MFCD00010903 CAS Number: 10043-52-4 EINECS: 233-140-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...