Ketanserin tartrate ≥98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-[2-[4-(4-Fluorobenzoyl)-1-piperdinyl)ethyl]-2,4(1H,3H)-quinazolinedione tartrate

J62798-MD
AAJ62798-MDEA 429 USD
AAJ62798-MD
Ketanserin tartrate ≥98%
Ketanserin tartrate

Selective 5-HT2A serotonin receptor antagonist


Powder


Applications: Selective 5-HT2A serotonin receptor antagonist

Formula: C₂₂H₂₂FN₃O₃·C₄H₆O₆
Storage Temperature: Ambient
MDL Number: MFCD00084651
CAS Number: 83846-83-7
EINECS: 281-062-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,5296

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR