EDTA (ethylenediamine tetraacetic acid) 0.5 M in aqueous solution pH 8.0
Supplier: Thermo Scientific Chemicals
|
Warning
|
J62786-AK
J62786-AP
AAJ62786-AKEA
128.02
USD
AAJ62786-AK
AAJ62786-AP
EDTA (ethylenediamine tetraacetic acid) 0.5 M in aqueous solution pH 8.0
EDTA (Ethylenediamine tetraacetic acid)
Liquid
CAS No.: 60-00-4
Molecular Formula: C10H16N2O8
Formula Weight: 292.24
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00003541
Formula:
C₁₀H₁₆N₂O₈ Storage Temperature: Ambient |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...