Fura 2-AM
Supplier: Thermo Scientific Chemicals
Synonyms:
FURA-2 pentakis(acetoxymethyl) ester, 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethyl ester, FURA-2 acetoxymethyl ester, Fura-2 AM Ester
A calcium indicator; derivative of Fura-2 used to load the chelator into cells
CAS No.: 108964-32-5
Molecular Formula: C44H47N3O24
Formula Weight: 1001.87
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Yellow
Beilstein Reference: 8183748
MDL No.: MFCD00036976
A calcium indicator; derivative of Fura-2 used to load the chelator into cells
Formula:
C₄₄H₄₇N₃O₂₄ MW: 1001.857 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00036976 CAS Number: 108964-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...