MES lysis buffer with Triton® X-100, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62693-AK
AAJ62693-AKEA
55
USD
AAJ62693-AK
AAJ62693-AP
MES lysis buffer with Triton® X-100, Thermo Scientific
Buffers
Extraction Buffers
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...