Mineral oil, white (petroleum), high purity
Supplier: Thermo Scientific Chemicals
Synonyms:
Matrix oil, Mineral oil, Paraffin oil
J62592-AE
J62592-AP
AAJ62592-AEEA
75.21
USD
AAJ62592-AE
AAJ62592-AP
Mineral oil, white (petroleum), high purity
White mineral oil (petroleum)
Suitable for PCR applications
Oily liquid
Applications: Suitable for PCR applications
Boiling Pt:
300 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00131611 CAS Number: 8042-47-5 EINECS: 232-455-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...