Sorbitan monooleate

Supplier: Thermo Scientific Chemicals

J62576-K2
AAJ62576-K2EA 152.09 USD
AAJ62576-K2
Sorbitan monooleate
Sorbitan monooleate

An emulsifier


Viscous liquid

Formula: C₂₄H₄₄O₆
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00080948
CAS Number: 1338-43-8
EINECS: 215-665-4
Merck Index: 14,8724

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR