L(-)-Tryptophan cell culture reagent
Supplier: Thermo Scientific Chemicals
Synonyms:
L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
J62508-09
J62508-22
J62508-A1
AAJ62508-09EA
32.1
USD
AAJ62508-09
AAJ62508-22
AAJ62508-A1
L(-)-Tryptophan cell culture reagent
L(-)-Tryptophan
Powder
CAS No.: 73-22-3
Molecular Formula: C11H12N2O2
Formula Weight: 204.23
Physical Form: Powder
Appearance: White to off-white
Melting Point: 280-285°
Beilstein Reference: 86197
Merck Reference: 14,9797
MDL No.: MFCD00064340
Formula:
C₁₁H₁₂N₂O₂ MW: 204.23 g/mol Melting Pt: 290 Storage Temperature: Ambient |
MDL Number:
MFCD00064340 CAS Number: 73-22-3 EINECS: 200-795-6 Merck Index: 14,9797 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...