Tetraethylene glycol monooctyl ether

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: n-Octyltetraoxyethylene, 3,6,9,12-Tetraoxaicosan-1-ol, 3,6,9,12-Tetraoxaeicosan-1-ol

J62496-06 J62496-03
AAJ62496-06EA 406 USD
AAJ62496-06 AAJ62496-03
Tetraethylene glycol monooctyl ether
Tetraethylene glycol monooctyl ether

Reagent for solubilizing membrane proteins


Solid


Applications: Reagent for solubilizing membrane proteins

Formula: C₁₆H₃₄O₅
MW: 306.443 g/mol
Density: 0.969
Storage Temperature: Freezer
MDL Number: MFCD00043033
CAS Number: 19327-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR